RIE156319654 Rieke Metals, Inc.

2-Ethoxy-1,1-difluorooxoethylzinc bromide

  • MDL number MFCD08064677
  • Cas Number : 109522-62-5
  • Molecular Weight : 268.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/2253

Safety Information

MSDS http://riekemetals.com/products-services/catnum/2253
Availability details for
Quantity
  • Availability
  • Add to Cart