RIE156320444 Rieke Metals, Inc.

2-Chloro-3'-iodobenzophenone

  • MDL number MFCD02260368
  • Cas Number : 890098-15-4
  • Molecular Weight : 342.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5078

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5078
Availability details for
Quantity
  • Availability
  • Add to Cart