RIE156320445 Rieke Metals, Inc.

3-chloro-3'-iodobenzophenone

  • MDL number MFCD02260369
  • Cas Number : 890098-16-5
  • Molecular Weight : 342.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5079

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5079
Availability details for
Quantity
  • Availability
  • Add to Cart