RIE156320451 Rieke Metals, Inc.

3-Chloro-4'-nitrobenzophenone

  • MDL number MFCD02260378
  • Cas Number : 131822-44-1
  • Molecular Weight : 261.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5090

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5090
Availability details for
Quantity
  • Availability
  • Add to Cart