RIE156320453 Rieke Metals, Inc.

3-fluoro-3'-nitrobenzophenone

  • MDL number MFCD02260383
  • Cas Number : 890098-21-2
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5096

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5096
Availability details for
Quantity
  • Availability
  • Add to Cart