RIE156320454 Rieke Metals, Inc.

3-fluoro-4'-nitrobenzophenone

  • MDL number MFCD02260386
  • Cas Number : 527744-61-2
  • Molecular Weight : 245.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-fluorophenyl)(4-nitrophenyl)methanone
Inchi Code 1S/C13H8FNO3/c14-11-3-1-2-10(8-11)13(16)9-4-6-12(7-5-9)15(17)18/h1-8H
Inchi Key XCFRMFKYPCTVCA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5099

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5099
Availability details for
Quantity
  • Availability
  • Add to Cart