RIE156320458 Rieke Metals, Inc.

3-fluoro-2'-iodobenzophenone

  • MDL number MFCD02260393
  • Cas Number : 890098-23-4
  • Molecular Weight : 326.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5109

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5109
Availability details for
Quantity
  • Availability
  • Add to Cart