RIE156320461 Rieke Metals, Inc.

3-fluoro-3'-iodobenzophenone

  • MDL number MFCD02260396
  • Cas Number : 890098-25-6
  • Molecular Weight : 326.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5112

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5112
Availability details for
Quantity
  • Availability
  • Add to Cart