RIE156320464 Rieke Metals, Inc.

3-fluoro-4'-iodobenzophenone

  • MDL number MFCD02260399
  • Cas Number : 890098-27-8
  • Molecular Weight : 326.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5115

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5115
Availability details for
Quantity
  • Availability
  • Add to Cart