RIE156320466 Rieke Metals, Inc.

2-Ethoxycarbonyl-3'-nitrobenzophenone

  • MDL number MFCD02260404
  • Cas Number : 890098-28-9
  • Molecular Weight : 299.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-(3-nitrobenzoyl)benzoate
Inchi Code 1S/C16H13NO5/c1-2-22-16(19)14-9-4-3-8-13(14)15(18)11-6-5-7-12(10-11)17(20)21/h3-10H,2H2,1H3
Inchi Key WMSZMJXCZHWMQV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5120

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5120
Availability details for
Quantity
  • Availability
  • Add to Cart