RIE156320468 Rieke Metals, Inc.

4-ethoxycarbonyl-3'-nitrobenzophenone

  • MDL number MFCD02260406
  • Cas Number : 890098-31-4
  • Molecular Weight : 299.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5122

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5122
Availability details for
Quantity
  • Availability
  • Add to Cart