RIE156320470 Rieke Metals, Inc.

3-Ethoxycarbonyl-4'-nitrobenzophenone

  • MDL number MFCD02260408
  • Cas Number : 760192-94-7
  • Molecular Weight : 299.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5124

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5124
Availability details for
Quantity
  • Availability
  • Add to Cart