RIE156320471 Rieke Metals, Inc.

2-ethoxycarbonyl-2'-iodobenzophenone

  • MDL number MFCD02260410
  • Cas Number : 890098-33-6
  • Molecular Weight : 380.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5126

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5126
Availability details for
Quantity
  • Availability
  • Add to Cart