RIE156320472 Rieke Metals, Inc.

3-ethoxycarbonyl-2'-iodobenzophenone

  • MDL number MFCD02260411
  • Cas Number : 890098-35-8
  • Molecular Weight : 380.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5127

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5127
Availability details for
Quantity
  • Availability
  • Add to Cart