RIE156320473 Rieke Metals, Inc.

4-Ethoxycarbonyl-2'-iodobenzophenone

  • MDL number MFCD02260412
  • Cas Number : 890098-37-0
  • Molecular Weight : 380.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5128

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5128
Availability details for
Quantity
  • Availability
  • Add to Cart