RIE156320484 Rieke Metals, Inc.

3-iodo-4'-thiomethylbenzophenone

  • MDL number MFCD02260426
  • Cas Number : 890098-59-6
  • Molecular Weight : 354.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5142

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5142
Availability details for
Quantity
  • Availability
  • Add to Cart