RIE156320485 Rieke Metals, Inc.

4-iodo-4'-thiomethylbenzophenone

  • MDL number MFCD02260427
  • Cas Number : 890098-61-0
  • Molecular Weight : 354.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-iodophenyl)[4-(methylsulfanyl)phenyl]methanone
Inchi Code 1S/C14H11IOS/c1-17-13-8-4-11(5-9-13)14(16)10-2-6-12(15)7-3-10/h2-9H,1H3
Inchi Key MPZJZWSFUJGBSO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5143

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5143
Availability details for
Quantity
  • Availability
  • Add to Cart