RIE156320491 Rieke Metals, Inc.

3-Cyano-4'-iodobenzophenone

  • MDL number MFCD02260433
  • Cas Number : 890098-73-4
  • Molecular Weight : 333.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5149

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5149
Availability details for
Quantity
  • Availability
  • Add to Cart