RIE156320492 Rieke Metals, Inc.

2-cyano-2'-iodobenzophenone

  • MDL number MFCD02260434
  • Cas Number : 890098-75-6
  • Molecular Weight : 333.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5150

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5150
Availability details for
Quantity
  • Availability
  • Add to Cart