RIE156320493 Rieke Metals, Inc.

2-cyano-3'-iodobenzophenone

  • MDL number MFCD02260435
  • Cas Number : 890098-77-8
  • Molecular Weight : 333.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5151

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5151
Availability details for
Quantity
  • Availability
  • Add to Cart