RIE156320494 Rieke Metals, Inc.

2-Cyano-4'-iodobenzophenone

  • MDL number MFCD02260436
  • Cas Number : 890098-79-0
  • Molecular Weight : 333.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-iodobenzoyl)benzonitrile
Inchi Code 1S/C14H8INO/c15-12-7-5-10(6-8-12)14(17)13-4-2-1-3-11(13)9-16/h1-8H
Inchi Key AKTKDGLWZCLYHY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5152

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5152
Availability details for
Quantity
  • Availability
  • Add to Cart