RIE156320495 Rieke Metals, Inc.

2-Methoxy-4'-nitrobenzophenone

  • MDL number MFCD03841017
  • Cas Number : 42495-50-1
  • Molecular Weight : 257.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5163

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5163
Availability details for
Quantity
  • Availability
  • Add to Cart