RIE156320497 Rieke Metals, Inc.

3-methyl-4'-nitrobenzophenone

  • MDL number MFCD00505992
  • Cas Number : 131822-45-2
  • Molecular Weight : 241.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5166

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5166
Availability details for
Quantity
  • Availability
  • Add to Cart