RIE156320498 Rieke Metals, Inc.

4-nitro-4'-thiomethylbenzophenone

  • MDL number MFCD07698795
  • Cas Number : 127615-48-9
  • Molecular Weight : 273.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5169

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5169
Availability details for
Quantity
  • Availability
  • Add to Cart