RIE156320499 Rieke Metals, Inc.

4-fluoro-3-methyl-4'-nitrobenzophenone

  • MDL number MFCD03841022
  • Cas Number : 760192-96-9
  • Molecular Weight : 259.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5170

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5170
Availability details for
Quantity
  • Availability
  • Add to Cart