RIE156320503 Rieke Metals, Inc.

2-Acetoxy-2'-methylbenzophenone

  • MDL number MFCD07698800
  • Cas Number : 890098-87-0
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-05
Availability details for
Quantity
  • Availability
  • Add to Cart