RIE156320505 Rieke Metals, Inc.

2-Acetoxy-4'-methylbenzophenone

  • MDL number MFCD07698802
  • Cas Number : 890098-91-6
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-07
Availability details for
Quantity
  • Availability
  • Add to Cart