RIE156320506 Rieke Metals, Inc.

2-Acetoxy-2'-trifluorobenzophenone

  • MDL number MFCD07698803
  • Cas Number : 890098-93-8
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-[2-(trifluoromethyl)benzoyl]phenyl acetate
Inchi Code 1S/C16H11F3O3/c1-10(20)22-14-9-5-3-7-12(14)15(21)11-6-2-4-8-13(11)16(17,18)19/h2-9H,1H3
Inchi Key XVJSAINOAGETFQ-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-08
Availability details for
Quantity
  • Availability
  • Add to Cart