RIE156320508 Rieke Metals, Inc.

2-Acetoxy-4'-trifluorobenzophenone

  • MDL number MFCD07698805
  • Cas Number : 890098-97-2
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-10
Availability details for
Quantity
  • Availability
  • Add to Cart