RIE156320509 Rieke Metals, Inc.

2-Acetoxy-2'-fluorobenzophenone

  • MDL number MFCD07698806
  • Cas Number : 890098-99-4
  • Molecular Weight : 258.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-11
Availability details for
Quantity
  • Availability
  • Add to Cart