RIE156320513 Rieke Metals, Inc.

2-Acetoxy-3'-chlorobenzophenone

  • MDL number MFCD07698810
  • Cas Number : 890099-10-2
  • Molecular Weight : 274.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-15
Availability details for
Quantity
  • Availability
  • Add to Cart