RIE156320515 Rieke Metals, Inc.

2-Acetoxy-2'-bromobenzophenone

  • MDL number MFCD07698812
  • Cas Number : 890099-16-8
  • Molecular Weight : 319.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-17

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-17
Availability details for
Quantity
  • Availability
  • Add to Cart