RIE156320524 Rieke Metals, Inc.

2-Acetoxy-4'-phenoxybenzophenone

  • MDL number MFCD07698821
  • Cas Number : 890098-30-3
  • Molecular Weight : 332.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(4-phenoxybenzoyl)phenyl acetate
Inchi Code 1S/C21H16O4/c1-15(22)24-20-10-6-5-9-19(20)21(23)16-11-13-18(14-12-16)25-17-7-3-2-4-8-17/h2-14H,1H3
Inchi Key JBZLVWLCVLNDKD-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-28
Availability details for
Quantity
  • Availability
  • Add to Cart