RIE156320539 Rieke Metals, Inc.

2-Acetoxy-4'-pentyloxybenzophenone

  • MDL number MFCD07698836
  • Cas Number : 890098-58-5
  • Molecular Weight : 326.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-36D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-36D
Availability details for
Quantity
  • Availability
  • Add to Cart