RIE156320545 Rieke Metals, Inc.

2-Acetoxy-2',6'-difluorobenzophenone

  • MDL number MFCD07698842
  • Cas Number : 890098-70-1
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-40
Availability details for
Quantity
  • Availability
  • Add to Cart