RIE156320546 Rieke Metals, Inc.

2-Acetoxy-3',4'-difluorobenzophenone

  • MDL number MFCD07698843
  • Cas Number : 890098-72-3
  • Molecular Weight : 276.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,4-difluorobenzoyl)phenyl acetate
Inchi Code 1S/C15H10F2O3/c1-9(18)20-14-5-3-2-4-11(14)15(19)10-6-7-12(16)13(17)8-10/h2-8H,1H3
Inchi Key KWGKYQWIEGMZMH-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-41
Availability details for
Quantity
  • Availability
  • Add to Cart