RIE156320548 Rieke Metals, Inc.

2-Acetoxy-2',3'-dichlorobenzophenone

  • MDL number MFCD07698845
  • Cas Number : 890098-76-7
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2,3-dichlorobenzoyl)phenyl acetate
Inchi Code 1S/C15H10Cl2O3/c1-9(18)20-13-8-3-2-5-10(13)15(19)11-6-4-7-12(16)14(11)17/h2-8H,1H3
Inchi Key UEVGKMFIPNOAAN-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-43
Availability details for
Quantity
  • Availability
  • Add to Cart