RIE156320550 Rieke Metals, Inc.

2-Acetoxy-2',5'-dichlorobenzophenone

  • MDL number MFCD07698847
  • Cas Number : 890098-80-3
  • Molecular Weight : 309.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-45
Availability details for
Quantity
  • Availability
  • Add to Cart