RIE156320554 Rieke Metals, Inc.

2-Acetoxy-2',3'-methoxybenzophenone

  • MDL number MFCD07698851
  • Cas Number : 890098-88-1
  • Molecular Weight : 300.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-49

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-49
Availability details for
Quantity
  • Availability
  • Add to Cart