RIE156320561 Rieke Metals, Inc.

2-Acetoxy-2',4'-methylbenzophenone

  • MDL number MFCD07698858
  • Cas Number : 890099-03-3
  • Molecular Weight : 268.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(2,4-dimethylbenzoyl)phenyl acetate
Inchi Code 1S/C17H16O3/c1-11-8-9-14(12(2)10-11)17(19)15-6-4-5-7-16(15)20-13(3)18/h4-10H,1-3H3
Inchi Key MHRCSKPNDZWJNO-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-56
Availability details for
Quantity
  • Availability
  • Add to Cart