RIE156320564 Rieke Metals, Inc.

2-Acetoxy-3',4'-methylbenzophenone

  • MDL number MFCD07698861
  • Cas Number : 890099-12-4
  • Molecular Weight : 268.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5175-59

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-59
Availability details for
Quantity
  • Availability
  • Add to Cart