RIE156320566 Rieke Metals, Inc.

2-Acetoxy-3',4',5'-trifluorobenzophenone

  • MDL number MFCD07698863
  • Cas Number : 890099-18-0
  • Molecular Weight : 294.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-(3,4,5-trifluorobenzoyl)phenyl acetate
Inchi Code 1S/C15H9F3O3/c1-8(19)21-13-5-3-2-4-10(13)15(20)9-6-11(16)14(18)12(17)7-9/h2-7H,1H3
Inchi Key YIJLYAUYWUPUNC-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5175-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5175-61
Availability details for
Quantity
  • Availability
  • Add to Cart