RIE156320573 Rieke Metals, Inc.

3-Acetoxy-4'-methylbenzophenone

  • MDL number MFCD07698871
  • Cas Number : 890099-36-2
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-07
Availability details for
Quantity
  • Availability
  • Add to Cart