RIE156320574 Rieke Metals, Inc.

3-Acetoxy-2'-trifluorobenzophenone

  • MDL number MFCD07698872
  • Cas Number : 890099-39-5
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-08
Availability details for
Quantity
  • Availability
  • Add to Cart