RIE156320575 Rieke Metals, Inc.

3-Acetoxy-3'-trifluorobenzophenone

  • MDL number MFCD07698873
  • Cas Number : 890099-41-9
  • Molecular Weight : 308.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-[3-(trifluoromethyl)benzoyl]phenyl acetate
Inchi Code 1S/C16H11F3O3/c1-10(20)22-14-7-3-5-12(9-14)15(21)11-4-2-6-13(8-11)16(17,18)19/h2-9H,1H3
Inchi Key IGJZQGRFNHTRNY-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5176-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-09
Availability details for
Quantity
  • Availability
  • Add to Cart