RIE156320578 Rieke Metals, Inc.

3-Acetoxy-3'-fluorobenzophenone

  • MDL number MFCD07698876
  • Cas Number : 890099-47-5
  • Molecular Weight : 258.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-12

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-12
Availability details for
Quantity
  • Availability
  • Add to Cart