RIE156320582 Rieke Metals, Inc.

3-Acetoxy-4'-chlorobenzophenone

  • MDL number MFCD07698880
  • Cas Number : 890099-55-5
  • Molecular Weight : 274.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-chlorobenzoyl)phenyl acetate
Inchi Code 1S/C15H11ClO3/c1-10(17)19-14-4-2-3-12(9-14)15(18)11-5-7-13(16)8-6-11/h2-9H,1H3
Inchi Key BHZTWVHDRPGENH-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5176-16

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-16
Availability details for
Quantity
  • Availability
  • Add to Cart