RIE156320584 Rieke Metals, Inc.

3-Acetoxy-3'-bromobenzophenone

  • MDL number MFCD07698882
  • Cas Number : 890099-59-9
  • Molecular Weight : 319.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-18

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-18
Availability details for
Quantity
  • Availability
  • Add to Cart