RIE156320593 Rieke Metals, Inc.

3-Acetoxy-3'-nitrobenzophenone

  • MDL number MFCD07698891
  • Cas Number : 890099-77-1
  • Molecular Weight : 285.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5176-30

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-30
Availability details for
Quantity
  • Availability
  • Add to Cart