RIE156320596 Rieke Metals, Inc.

3-Acetoxy-4'-isopropoxybenzophenone

  • MDL number MFCD07698894
  • Cas Number : 890099-84-0
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(4-isopropoxybenzoyl)phenyl acetate
Inchi Code 1S/C18H18O4/c1-12(2)21-16-9-7-14(8-10-16)18(20)15-5-4-6-17(11-15)22-13(3)19/h4-12H,1-3H3
Inchi Key CEPSSVOKNOCGSN-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5176-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5176-33
Availability details for
Quantity
  • Availability
  • Add to Cart